missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CoA Synthase Recombinant Rabbit Monoclonal Antibody (23GB3095), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553317
This item is not returnable.
View return policy
Description
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. COASY is a bifunctional enzyme that catalyzes the 2 last steps in CoA synthesis. These activities are performed by 2 separate enzymes, phosphopantetheine adenylyltransferase and dephospho-CoA kinase, in prokaryotes.
Specifications
| CoA Synthase | |
| Recombinant Monoclonal | |
| Unconjugated | |
| COASY | |
| 1300003G02Rik; bifunctional coenzyme A synthase; bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase; CoA synthase; Coasy; Coenzyme A synthase; Dephospho-CoA kinase; Dephospho-CoA pyrophosphorylase; Dephosphocoenzyme A kinase; DPCK; DPCOAK; NBIA6; NBP; nucleotide binding protein; Pantetheine-phosphate adenylyltransferase; Phosphopantetheine adenylyltransferase; phosphopantetheine adenylyltransferase / dephosphocoenzyme A kinase; POV-2; PPAT; PSEC0106; Ukr1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 80347 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3095 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13057 | |
| COASY | |
| A synthesized peptide derived from COASY (530-564AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction