Learn More
Invitrogen™ CoA Synthase Monoclonal Antibody (OTI3B3)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA525654
Product Code. 15945254
  • £474.00 / 100µL
Description
CoA Synthase Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. COASY is a bifunctional enzyme that catalyzes the 2 last steps in CoA synthesis. These activities are performed by 2 separate enzymes, phosphopantetheine adenylyltransferase and dephospho-CoA kinase, in prokaryotes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CoA Synthase
Monoclonal
0.75 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q13057
COASY
Full length human recombinant protein of COASY produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI3B3
Unconjugated
COASY
1300003G02Rik; bifunctional coenzyme A synthase; bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase; CoA synthase; Coasy; Coenzyme A synthase; Dephospho-CoA kinase; Dephospho-CoA pyrophosphorylase; Dephosphocoenzyme A kinase; DPCK; DPCOAK; NBIA6; NBP; nucleotide binding protein; Pantetheine-phosphate adenylyltransferase; Phosphopantetheine adenylyltransferase; phosphopantetheine adenylyltransferase / dephosphocoenzyme A kinase; POV-2; PPAT; PSEC0106; Ukr1
Mouse
Affinity Chromatography
RUO
80347
-20°C, Avoid Freeze/Thaw Cycles