missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ CO2 Incubator Series CBF

Product Code. 31741125 Shop All BINDER Products
Change view
Click to view available options
Dimensions (D x W x H) Interior:
500 x 560 x 600 mm
575 x 620 x 750 mm
No. of Shelves:
3 std./6 max.
3 std./8 max.
Model:
CBF170
CBF170-O
CBF260
CBF260-O
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (D x W x H) Interior No. of Shelves Model unitSize
31741125 575 x 620 x 750 mm 3 std./8 max. CBF260 Each
31741148 500 x 560 x 600 mm 3 std./6 max. CBF170 Each
31741135 500 x 560 x 600 mm 3 std./6 max. CBF170-O Each
31741119 575 x 620 x 750 mm 3 std./8 max. CBF260-O Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31741125 Supplier BINDER™ Supplier No. 90400215

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Series CBF - CO2 incubators with hot air sterilization and humidity regulation.

The BINDER CBF is a high-end CO2 incubator with humidity control, suitable for all sensitive incubation applications in GMP environments. It ensures consistent results, including in cell-based assays with multiwell plates. This BINDER incubator is even suitable for complex cultivation experiments or individual environments with hypoxic conditions, thanks to its comprehensive assortment of options and accessories.

  • Safe - cell cultures receive maximum protection against contamination
  • Reliable - constant well-being conditions for the cells
  • Smart - simple routine cleaning and convenient operation
  • Economical - efficient operation without consumables

IMPORTANT FEATURES

  • Contamination-safe humidification system without water pan
  • Tried-and-tested anti-contamination concept with 180°C hot air sterilization
  • Hot-air sterilizable CO2 sensor with single-beam infrared technology
  • Seamless inner chamber made of stainless steel with flanges as shelf support system
  • Fanless interior with Venturi CO2 gas mixing nozzle
  • Intuitive touchscreen controller
  • Internal data logger, measured values can be read out in open format via USB
  • Troubleshooting system with visual and acoustic alarms
  • Interfaces: Ethernet, USB, zero-voltage alarm contact
  • Fail-safe CO2 system for protecting the pH of cell cultures

OPTIONAL EQUIPMENT

  • O2 control with O2 control ranges: 0.2 to 20 vol.% O2 or 10 to 90 vol.% O2
  • Multi-sectional glass doors with special shelves for dividing up the space to accommodate different experiments
  • Stacking adapter for secure, space-saving installation
  • Gas tank changer for connecting two gas tanks to one or two incubators

Specifications

Controller Type MB 2
Capacity (Metric) 267 L
Chamber Material Stainless Steel (304/1.4301)
CO2 Concentration Range 0 to 20%
CO2 Sensor Technology IR Sensor
Convection Type Natural
Material Stainless Steel
No. of Places 110
No. of Shelves 3 std./8 max.
Plug Type EU
Relative Humidity 50 to 95%
Temperature Range (Metric) +6 above ambient to 50°C
Type CO2 Incubator
Display Yes
Model CBF260
Temperature Uniformity 0.4 ± K
Dimensions (D x W x H) Interior 575 x 620 x 750 mm
Height (Metric) 1020 mm
Depth (Metric) Interior 575 mm
Weight (Metric) 126 kg
Voltage 200/230 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.