Learn More
Thermo Scientific™ CO2 and LN2 Backup Systems
Product Code. p-4530851 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Thermo Scientific Cryogenic Freezers
Type:
LN2 Backup System
Unit Size:
Each
Product Code. 10552111
Brand: Thermo Scientific™ 6595
Only null left
Add to basket
Add to basket
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems with ultra low-temperature freezers.
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems with ultra low-temperature freezers. Systems inject liquid CO2 or LN2 into cabinet when cabinet temperature warms to preset level.
Features
  • Temperature is adjustable in 1°C increments to -75°C (CO2), -150°C (LN2).
  • Digital display constantly indicates actual cabinet temperature.
  • Indicator lights come on when system is activated.
  • Additional indicator lights when cylinder is empty.
  • Liquid flow is shut off automatically when freezer door or lid is opened.
  • System is powered by rechargeable battery.
  • Low-battery light indicates when battery requires charging.
Ordering Information:
Thermo Scientific Revco™ Ultima™ II and Elite™ 3.1 cu. ft. (88L) models and all Value series models require freestanding system; only Ultima II and Elite series models larger than 3.1 cu. ft. (88L) accept built-in systems. Specify freezer model and voltage when ordering.
Note:
Customer-installed units; must be installed by qualified professional.
Includes:
All required hardware except CO2 or LN2 cylinder (must be ordered separately)
TRUSTED_SUSTAINABILITY
Specifications
Type LN2 Backup System
Capacity (English) 3.1 cu. ft.
Capacity (Metric) 88 L
Temperature Range -150°C to +1°C
Style Built-in
For Use With (Equipment) Thermo Scientific Cryogenic Freezers
Defrost Automatic