missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ CO2 Incubator ICOmed (Medical Device)

Product Code. 15851575 Shop All Memmert Products
Click to view available options
Capacity (Metric):
107 L
156 L
241 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15851575

Brand: Memmert™ ICO150med

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for the creation and maintenance of constant environmental conditions for application in the field of in vitro fertilisation (IVF), especially for the incubation of oocytes, spermatozoa and zygotes in special culture dishes for IVF application.

Sterile, safe and reliable – these three words describe the Memmert CO2 incubator ICOmed for the medical use.
Memmert subjected its CO2 incubator ICOmed to a comprehensive medical device evaluation. Each Memmert CO2 incubator ICOmed is classified as a Class IIa medical device for the in vitro fertilisation (IVF) as well as biosynthesis.

  • Model sizes: 50, 105, 150, 240
  • Temperature adjustment range: +18 °C to +50 °C
  • CO2 adjustment range: 0 to 20 %
  • Humidity adjustment ranges: 40 to 97 % rh (with option K7)
  • O2 adjustment ranges: 1 to 20 % (with option T6)
  • Classified as a medical device class IIa
  • 1 model variant: TwinDISPLAY (intuitive control)
  • Perfect distribution of temperature, humidity and CO2
  • Long-time stable and repetitively accurate
  • The interior chamber, including all installations and sensors, can be sterilised at +180 °C in a 60-minute programme
  • Visual and acoustic alarm as well as alarm notification to mobile phones if the individually adjustable temperature, CO2, O2, or humidity ranges are exceeded
  • Battery-buffered ControlCOCKPIT: The operating display, logging and CO2 control are fully functional even during a power failure
  • Fail-safe, FDA-compliant logging of temperature, CO2, O2 humidity, open door time data
  • Adjustable parameters: temperature (Celsius or Fahrenheit), CO2, programme time, time zones, summertime/wintertime

Active humidity control ensures short recovery times after the door has been opened and minimises vaporisation in the interior. Together with the heating of the interior from all six sides and the heated inner glass door, it offers maximum protection for cell and tissue cultures and avoids the dangerous formation of condensation. The turbulence-free chamber ventilation ensures a constant and uniform atmosphere.

Specifications

Alarm Types Optic and acoustic
Capacity (Metric) Shelf 15 kg
Certifications/Compliance DIN 12880:2007-05 , EN 61010-1 (IEC 61010-1), EN 61010-2-010
Controller Type TwinDISPLAY: adaptive multifunctional digital PID-microprocessor controller with 2 high-definition TFT-colour displays
Capacity (Metric) 156 L
Chamber Material Stainless Steel
CO2 Concentration Range 0 to 20%
CO2 Sensor Technology IR Sensor
Material Textured Stainless Steel
No. of Shelves 2
O2 Concentration Range 1 to 20%
Relative Humidity 40 to 97%
Temperature Range (Metric) 18°C to 50°C
Type CO2 Incubator
Disinfection Time 60 min.
Display 2 high-definition TFT-colour displays (TwinDISPLAY)
Ports Mains cable with plug (German type)
Shelf Material Perforated Stainless Steel
Dimensions (D x W x H) Interior 400 x 560 x 700 mm
Dimensions (L x W x H) 591 x 719 x 1066 mm
Shipping Weight (Metric) 116 kg
Weight (Metric) 90 kg
Electrical Requirements 115/230 V, 50/60 Hz
Frequency 50/60 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.