missing translation for 'onlineSavingsMsg'
Learn More

CNX-2006, MedChemExpress

Product Code. 16475898
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16475898 10 mg 10mg
16462047 50 mg 50mg
16458537 5 mg 5mg
16406356 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16475898 Supplier Medchem Express Supplier No. HY13897
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CNX-2006 is a mutant-selective and irreversible EGFR inhibitor with an IC50 below 20 nM for EGFRT790M.

CNX-2006 is a mutant-selective and irreversible EGFR inhibitor with an IC50 below 20 nM for EGFRT790M.

Specifications

Chemical Name or Material CNX-2006
CAS 1375465-09-0
Color Gray
Purity Grade Notes Research
Molecular Formula C26H27F4N7O2
Quantity 10 mg
Solubility Information DMSO : ≥ 52 mg/mL (95.32 mM)
SMILES C=CC(NC1=CC=CC(NC2=NC(NC3=CC=C(NC4CN(CCF)C4)C=C3OC)=NC=C2C(F)(F)F)=C1)=O
Molecular Weight (g/mol) 545.53
Formula Weight 545.53
Percent Purity 99.68%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.