missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNTROB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554760
This item is not returnable.
View return policy
Description
Immunogen sequence: LEELFPRYTS LRPGPPLNPP DFQGLRDALD SEHTRRKHCE RHIQSLQTRV LELQQQLAVA VAADRKKDTM IEQLDKTLAR VVEGWNRHEA ERTEVLRGLQ EEHQAAELTR SKQQE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
CNTROB is a centrosomal protein that interacts with BRCA2 and is required for centriole duplication and cytokinesis (Zou et al., 2005 ).
Specifications
| CNTROB | |
| Polyclonal | |
| Unconjugated | |
| CNTROB | |
| 9830165K03Rik; centrobin; centrobin, centriole duplication and spindle assembly protein; centrobin, centrosomal BRCA2 interacting protein; centrosomal BRCA2-interacting protein; CNTROB; LIP8; LYST-interacting protein 8; LYST-interacting protein LIP8; PP1221; RGD1307488 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116840 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N137 | |
| CNTROB | |
| Recombinant protein corresponding to Human CNTROB. Recombinant protein control fragment (Product #RP-93414). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction