missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNTNAP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103766
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CNTNAP4.
Presynaptic protein involved in both dopaminergic synaptic transmission and GABAergic system, thereby participating in the structural maturation of inhibitory interneuron synapses. Involved in the dopaminergic synaptic transmission by attenuating dopamine release through a presynaptic mechanism. Also participates in the GABAergic system.
Specifications
| CNTNAP4 | |
| Polyclonal | |
| Unconjugated | |
| CNTNAP4 | |
| Caspr4; cell recognition molecule Caspr4; cell recognition protein CASPR4; Cntnap4; contactin associated protein 4; contactin associated protein like 4; contactin associated protein-like 4; contactin-associated protein-like 4; E130114F09Rik; KIAA1763 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 170571, 307865, 85445 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q99P47, Q9C0A0 | |
| CNTNAP4 | |
| A synthesized peptide derived from human CNTNAP4(Accession Q9C0A0), corresponding to amino acid residues S1201-I1251. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction