Learn More
Invitrogen™ CNTNAP1 Recombinant Rabbit Monoclonal Antibody (HL3160)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556623
Product Code. 30276142
  • £434.00 / 100µL
Description
CNTNAP1 Recombinant Monoclonal Antibody for Western Blot
CNTNAP1 was initially identified as a 190-kDa protein associated with the contactin-PTPRZ1 complex. The 1, 384-amino acid protein, also designated p190 or CASPR for 'contactin-associated protein, ' includes an extracellular domain with several putative protein-protein interaction domains, a putative transmembrane domain, and a 74-amino acid cytoplasmic domain. The architecture of its extracellular domain is similar to that of neurexins, and this protein may be the signaling subunit of contactin, enabling recruitment and activation of intracellular signaling pathways in neurons. The gene product was initially identified as a 190-kDa protein associated with the contactin-PTPRZ1 complex. The 1, 381-amino acid protein, also designated p190 or CASPR for 'contactin-associated protein, ' includes an extracellular domain with several putative protein-protein interaction domains, a putative transmembrane domain, and a 74-amino acid cytoplasmic domain. Northern blot analysis showed that the gene is transcribed predominantly in brain as a transcript of 6.2 kb, with weak expression in several other tissues tested. The architecture of its extracellular domain is similar to that of neurexins, and this protein may be the signaling subunit of contactin, enabling recruitment and activation of intracellular signaling pathways in neurons.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CNTNAP1
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
O54991, P78357, P97846
Cntnap1
The immunogen used to generate this antibody corresponds to human CASPR.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot
HL3160
Unconjugated
Cntnap1
AI841080; Caspr; caspr1; CNTNAP; CNTNAP1; contactin associated protein 1; contactin associated protein-like 1; contactin-associated protein 1; LOW QUALITY PROTEIN: contactin-associated protein 1; MHDNIV; NCP1; neurexin 4; neurexin 4 (contactin associated protein); neurexin IV; Neurexin-4; NRXN4; p190; paranodin; shm
Rabbit
Protein A
RUO
53321, 84008, 8506
-20°C, Avoid Freeze/Thaw Cycles