missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNTNAP1 Recombinant Rabbit Monoclonal Antibody (23GB5605), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CNTNAP1 |
|---|---|
| Clone | 23GB5605 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088751
![]() |
Invitrogen™
MA553910 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088453
|
Invitrogen™
MA553911 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CNTNAP1 Recombinant Monoclonal antibody specifically detects CNTNAP1 in Human, Mouse samples. It is validated for Flow Cytometry, Western BlotSpecifications
| CNTNAP1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| O54991, P78357 | |
| 53321, 8506 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5605 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI841080; Caspr; caspr1; CNTNAP; CNTNAP1; contactin associated protein 1; contactin associated protein-like 1; contactin-associated protein 1; LOW QUALITY PROTEIN: contactin-associated protein 1; MHDNIV; NCP1; neurexin 4; neurexin 4 (contactin associated protein); neurexin IV; Neurexin-4; NRXN4; p190; paranodin; shm | |
| A synthesized peptide derived from human Caspr (700-770AA). | |
| Primary | |
| -20°C | |
| Cntnap1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title