Learn More
CNTN2 Mouse anti-Human, Unconjugated, Clone: 1D3G5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67089-1-IG-150UL
This item is not returnable.
View return policy
Description
The CNTN2 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human CNTN2. This antibody recognizes human, pig, mouse antigen. The CNTN2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| CNTN2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Pig |
| Western Blot, Immunohistochemistry, ELISA | |
| 1D3G5 | |
| Unconjugated | |
| 150 μL | |
| 6900 | |
| IgG2b |