missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNTN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109564
This item is not returnable.
View return policy
Description
Immunogen sequence: LECFALGNPV PDIRWRKVLE PMPSTAEIST SGAVLKIFNI QLEDEGIYEC EAENIRGKDK HQARIYVQAF PEWVEHINDT EVDIGSDLYW PCVATGKPIP T.
Contactin 1 encodes a protein that is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| CNTN1 | |
| Polyclonal | |
| Unconjugated | |
| Cntn1 | |
| AW495098; CNTN; CNTN1; contactin 1; contactin-1; F3; F3cam; glycoprotein gP135; GP135; MYPCN; neural cell surface protein F3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 117258, 1272, 12805 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.94 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P12960, Q12860, Q63198 | |
| Cntn1 | |
| A synthetic peptide corresponding to a sequence within amino acids 250-350 of human CNTN1 (NP_778203.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction