missing translation for 'onlineSavingsMsg'
Learn More

CNTFR Antibody, Novus Biologicals™

Product Code. 18252872 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18252872 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18252872 Supplier Novus Biologicals Supplier No. NBP168943

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CNTFR Polyclonal specifically detects CNTFR in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNTFR
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P26992
Gene Alias ciliary neurotrophic factor receptor, ciliary neurotrophic factor receptor alpha, ciliary neurotrophic factor receptor subunit alpha, CNTF receptor subunit alpha, CNTFR alpha, CNTFR-alpha, MGC1774
Gene Symbols CNTFR
Host Species Rabbit
Immunogen Synthetic peptides corresponding to CNTFR (ciliary neurotrophic factor receptor) The peptide sequence was selected from the C terminal of CNTFR. Peptide sequence VAAHATPWTEEPRHLTTEAQAAETTTSTTSSLAPPPTTKICDPGELGSGG.
Molecular Weight of Antigen 41 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1271
Test Specificity This product is specific to Subunit or Isoform: alpha.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.