Learn More
CNTF Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 500-P140BT-1MG
Description
AA Sequence of recombinant protein: AFTEHSPLTP HRRDLCSRSI WLARKIRSDL TALTQSYVKH QGLNKNINLD SADGMPVAST DQWSQLTQAQ RLQQNLQAYR TFHVLLARLL QDQQVHFTPT QGDFHQAIHT LLLQVAAFAY QIQQLMILLQ YKIPRNQADG MPINVGDGGL FQKKLWGLKV LQQLSQWTVR SIHDLRFISS HQTGIPARGS HYIANNKKM. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human CNTF. Anti-Human CNTF-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human CNTF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human CNTF (500-P140) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human CNTF. Western Blot: To detect hCNTF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hCNTF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P140BT-1MG will be provided as 2 x 500 μg
The protein encoded by this gene is a polypeptide hormone whose actions appear to be restricted to the nervous system where it promotes neurotransmitter synthesis and neurite outgrowth in certain neuronal populations. The protein is a potent survival factor for neurons and oligodendrocytes and may be relevant in reducing tissue destruction during inflammatory attacks. A mutation in this gene, which results in aberrant splicing, leads to ciliary neurotrophic factor deficiency, but this phenotype is not causally related to neurologic disease. A read-through transcript variant composed of ZFP91 and CNTF sequence has been identified, but it is thought to be non-coding. Read-through transcription of ZFP91 and CNTF has also been observed in mouse.
Specifications
| CNTF | |
| Polyclonal | |
| Biotin | |
| CNTF | |
| AI429687; Ciliary neurotrophic factor; ciliary neurotropic factor; Cntf; HCNTF; H-CNTF | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1270 | |
| -20°C |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P26441 | |
| CNTF | |
| E.coli-derived Recombinant Human CNTF | |
| 2 x 500 μg | |
| Primary | |
| Human | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.