missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNTF Monoclonal Antibody (21809)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523710
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
The protein encoded by this gene is a polypeptide hormone whose actions appear to be restricted to the nervous system where it promotes neurotransmitter synthesis and neurite outgrowth in certain neuronal populations. The protein is a potent survival factor for neurons and oligodendrocytes and may be relevant in reducing tissue destruction during inflammatory attacks. A mutation in this gene, which results in aberrant splicing, leads to ciliary neurotrophic factor deficiency, but this phenotype is not causally related to neurologic disease. A read-through transcript variant composed of ZFP91 and CNTF sequence has been identified, but it is thought to be non-coding. Read-through transcription of ZFP91 and CNTF has also been observed in mouse.
Specifications
| CNTF | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P26441 | |
| CNTF | |
| E. coli-derived recombinant human CTNF Ala2-Met200. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Neutralization, Western Blot | |
| 21809 | |
| Unconjugated | |
| CNTF | |
| AI429687; Ciliary neurotrophic factor; ciliary neurotropic factor; Cntf; HCNTF; H-CNTF | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1270 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction