missing translation for 'onlineSavingsMsg'
Learn More

CNTF Antibody (001), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18420489 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
18420489 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420489 Supplier Novus Biologicals Supplier No. NBP289998MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CNTF Monoclonal antibody specifically detects CNTF in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Dilution ELISA
Formulation 50mM Sodium Borate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.