missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTF Antibody (001), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89998DL594
This item is not returnable.
View return policy
Description
CNTF Monoclonal specifically detects CNTF in Human samples. It is validated for ELISA.
Specifications
| CNTF | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 1 | |
| ELISA | |
| ciliary neurotrophic factor, HCNTF | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CNTF (NP_000605.1; Ala 2-Met 200). | |
| 0.1 mL | |
| Biologically Active Proteins, Neuroscience | |
| 1270 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction