missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA561049
This item is not returnable.
View return policy
Description
Immunogen sequence: SRLGPIVRRW APRPSPLQSL AASLDAEPSS AAVPDGFPAG PTVSPRRLAR PPGLEEALSA LGLQGEREYA GDIFAEVM Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 33%.
CNTD2 gene ontology annotations related to this gene include mitotic cell cycle; positive regulation of cell cycle; positive regulation of cell proliferation; protein phosphorylation; regulation of cyclin-dependent protein serine/threonine kinase activity; regulation of mitotic nuclear division.
Specifications
| CNTD2 | |
| Polyclonal | |
| Unconjugated | |
| CNTD2 | |
| CCNP; CNTD2; cyclin N-terminal domain containing 2; cyclin N-terminal domain-containing protein 2; cyclin P; LOW QUALITY PROTEIN: cyclin N-terminal domain-containing protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79935 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H8S5 | |
| CNTD2 | |
| Recombinant protein corresponding to Human CNTD2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction