missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTD Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92384-0.1ml
This item is not returnable.
View return policy
Description
CNTD Polyclonal antibody specifically detects CNTD in Human samples. It is validated for Western Blot
Specifications
| CNTD | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CNTD, cyclin N-terminal domain containing, cyclin N-terminal domain containing 1, cyclin N-terminal domain-containing protein 1, FLJ40137 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human CNTD1 (NP_001317151.1). MVKQAENICRQATIQPRDNKRESQNWRALKQQLVNKFTLRLVSCVQLASKLSFRNKIISNITVLNFLQALGYLHTKEELLESELDVLKSLNFRINLPTPLAYVETLLEVLGYNGCLVPAM | |
| 0.1 mL | |
| Cell Biology | |
| 124817 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction