missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNST Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552405
This item is not returnable.
View return policy
Description
Immunogen sequence: PKEGGATTKE SESKTCLGTE SSKESQHTVE PLGSSPCCHQ MDVQTDSPSL SVTAGKDHME ELLCSAEATL ALHTQSSETA GSPSGPDSSE DACEDDSRLQ LAQTEACQDV ARIEGIAEDP KVFLSSKSKT EPLISPGCDR IPPAL Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 49%.
Required for targeting of connexins to the plasma membrane.
Specifications
| CNST | |
| Polyclonal | |
| Unconjugated | |
| CNST | |
| 9630058J23Rik; C1orf71; CNST; Consortin; consortin, connexin sorting protein; PPP1R64; protein phosphatase 1, regulatory subunit 64 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 163882 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6PJW8 | |
| CNST | |
| Recombinant protein corresponding to Human CNST. Recombinant protein control fragment (Product #RP-89727). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction