missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNST Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06530PCP
This item is not returnable.
View return policy
Description
CNST Polyclonal antibody specifically detects CNST in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CNST | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| C1orf71, consortin, connexin sorting protein, FLJ32001, MGC18089, PPP1R64, protein phosphatase 1, regulatory subunit 64 | |
| Produced in rabbits immunized with E. coli-derived Human CNST fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 163882 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction