missing translation for 'onlineSavingsMsg'
Learn More

CNRIP1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18420046 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420046 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420046 Supplier Novus Biologicals Supplier No. NBP281897AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CNRIP1 Polyclonal antibody specifically detects CNRIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNRIP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias C2orf32, cannabinoid receptor CB1-interacting protein 1, cannabinoid receptor interacting protein 1, CB1 cannabinoid receptor-interacting protein 1, chromosome 2 open reading frame 32, CRIP1, CRIP-1, CRIP1a, CRIP1b, DKFZp566K1924
Host Species Rabbit
Immunogen CNRIP1 antibody was raised against a 15 amino acid peptide near the center of human CNRIP1. The immunogen is located within amino acids 50 - 100 of CNRIP1. Amino Acid Squence: VPLELKSKEPDGDRV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 25927
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.