missing translation for 'onlineSavingsMsg'
Learn More

CNQX, MedChemExpress

Product Code. 16406308
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16406308

Brand: Medchem Express HY15066

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CNQX (FG9065) is a potent and competitive AMPA/kainate receptor antagonist with IC50s of 0.3 μM and 1.5 μM, respectively. CNQX is a competitive non-NMDA receptor antagonist. CNQX blocks the expression of fear-potentiated startle in rats.

CNQX (FG9065) is a potent and competitive AMPA/kainate receptor antagonist with IC50s of 0.3 μM and 1.5 μM, respectively. CNQX is a competitive non-NMDA receptor antagonist. CNQX blocks the expression of fear-potentiated startle in rats.

Specifications

Chemical Name or Material CNQX
Physical Form Solid
CAS 115066-14-3
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C9H4N4O4
For Use With (Application) Neuroscience-Neurodegeneration
Synonym FG9065
Solubility Information DMSO : 20 mg/mL (86.15 mM; Need ultrasonic)
SMILES O=C1C(NC2=CC([N+]([O-])=O)=C(C#N)C=C2N1)=O
Molecular Weight (g/mol) 232.15
Formula Weight 232.15
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.