missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNPY3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15215-1-AP-150UL
This item is not returnable.
View return policy
Description
PRAT4A is a novel protein belonging to the canopy family with a saposin B-type domain. It is an accessory molecule required for microbial recognition by Toll-like receptor (TLR), and mediates TLR dependent immune responses. Though the exact function of PRAT4A is still unknown, reports suggest that it is associated with the immature form of TLR4 and regulates the cell surface expression of this TLR. PRAT4A, a secreted protein, also has a role in TLR4 maturation and glycosylation. PRATA4 is usually present in endoplasmic reticulum and plays an active role in regulating the subcellular distribution and response of multiple TLRs and is required for both innate and adaptive immune responses.Specifications
| CNPY3 | |
| Polyclonal | |
| Unconjugated | |
| CNPY3 | |
| CAG4A, canopy 3 homolog (zebrafish), CNPY3, CTG repeat protein 4a, CTG4A, ERDA5, PRAT4A, Protein associated with TLR4, Protein canopy homolog 3, TNRC5, UNQ1934/PRO4409 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10695, 685174, 72029 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BT09, Q9DAU1 | |
| Cnpy3 | |
| CNPY3 Fusion Protein Ag7382 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction