missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNPY2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558403
This item is not returnable.
View return policy
Description
Immunogen sequence: RRSQDLHCGA CRALVDELEW EIAQVDPKKT IQMGSFRINP DGSQSVVEVP YARSEAHLTE LLEEICDRMK EYGEQIDPST HRKNYVRVVG RNGESSELDL QGIRIDSD Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
CNPY2 is positive regulator of neurite outgrowth by stabilizing myosin regulatory light chain (MRLC). It prevents MIR-mediated MRLC ubiquitination and its subsequent proteasomal degradation.
Specifications
| CNPY2 | |
| Polyclonal | |
| Unconjugated | |
| CNPY2 | |
| 5330432A10Rik; AW229003; canopy 2 homolog; canopy 2 homolog (zebrafish); canopy FGF signaling regulator 2; Cnpy2; D10Bwg1546e; HP10390; MIR-interacting saposin-like protein; MSAP; Protein canopy homolog 2; putative secreted protein Zsig9; Tmem4; transmembrane protein 4; UNQ1943/PRO4426; Zsig9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10330 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2B0 | |
| CNPY2 | |
| Recombinant protein corresponding to Human CNPY2. Recombinant protein control fragment (Product #RP-97779). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction