Learn More
CNPY2, MSAP Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-14635-100UL
Description
The CNPY2, MSAP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CNPY2, MSAP. This antibody recognizes human, mouse, rat antigen. The CNPY2, MSAP antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| CNPY2, MSAP | |
| 1000 μg/mL | |
| canopy 2 homolog (zebrafish), CNPY2, HP10390, MSAP, Protein canopy homolog 2, TMEM4, Transmembrane protein 4, ZSIG9 | |
| 100 μL | |
| 10330 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |