missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CNPase Polyclonal Antibody

Product Code. 17816587
Change view
Click to view available options
Quantity:
100 μL
500 μL
Unit Size:
100µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17816587 500 μL 500µL
17886697 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17816587 Supplier Invitrogen™ Supplier No. PA5143530

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chicken Polyclonal Antibody

CNPase Polyclonal Antibody for ICC/IF, IHC (P), IHC (PFA), Western Blot

CNP may participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNPase
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CNP
Gene Accession No. P06623, P09543, P13233, P16330
Gene Alias 2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; CNP; Cnp1; Cnp-1; CNPase; CNPF; CNPI; CNPII; cyclic nucleotide phosphodiesterase 1
Gene Symbols CNP
Host Species Chicken
Immunogen Full length human recombinant protein.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100066401, 100511587, 1267, 12799, 25275, 280752
Target Species Bovine, Horse, Human, Mouse, Pig, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.