missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNPase Mouse anti-Human, Porcine, Rat, Clone: 1B9G9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66729-1-IG-20UL
This item is not returnable.
View return policy
Description
CNP may participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin.Specifications
| CNPase | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P09543, P13233 | |
| CNP | |
| CNPase Fusion Protein Ag4227 | |
| 20 μL | |
| Primary | |
| Pig, Human, Rat | |
| Antibody | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1B9G9 | |
| Unconjugated | |
| CNP | |
| CNP, CNP1, CNPase | |
| Mouse | |
| Protein G | |
| RUO | |
| 100511587, 1267, 25275 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction