missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNPase Antibody (CL2872) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46636C
This item is not returnable.
View return policy
Description
CNPase Monoclonal antibody specifically detects CNPase in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| CNPase | |
| Monoclonal | |
| DyLight 650 | |
| 2', 3' cyclic nucleotide 3' phosphohydrolase, 2'-3'-cyclic nucleotide 3' phosphodiesterase, 2'-3'-cyclic-nucleotide 3'-phosphodiesterase, CNP1, CNPase, EC 3.1.4.37 | |
| This antibody was developed using a recombinant protein derived from P09543, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cellular Markers, Glia Markers, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers | |
| 1267 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL2872 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction