missing translation for 'onlineSavingsMsg'
Learn More

CNP/NPPC Antibody (347608) [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30735942 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735942 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735942 Supplier Novus Biologicals Supplier No. FAB3127MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CNP/NPPC Monoclonal antibody specifically detects CNP/NPPC in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNP/NPPC
Applications Western Blot
Classification Monoclonal
Clone 347608
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias CNP, CNP2, C-type natriuretic peptide, natriuretic peptide C, natriuretic peptide precursor C
Host Species Rat
Immunogen E. coli-derived recombinant human CNP, Lys24-Cys126, Accession # P23582
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 4880
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.