missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOX Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92146-0.1ml
This item is not returnable.
View return policy
Description
CNOX Polyclonal antibody specifically detects CNOX in Human, Mouse samples. It is validated for Western Blot
Specifications
| CNOX | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| candidate growth-related and time keeping constitutive hydroquinone (NADH)oxidase, Candidate growth-related and time keeping constitutive hydroquinone [NADH]oxidase, cCNOXConstitutive Ecto-NOX, Cell proliferation-inducing gene 38 protein, cNOX, CNOXbA64J21.1, ecto-NOX disulfide-thiol exchanger 1, FLJ10094, PIG38, proliferation-inducing protein 38 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-560 of human ENOX1 (NP_060463.2). LKQEKEQLFRTEENLTKDQQLQFLQQTMQGMQQQLLTIQEELNNKKSELEQAKEEQSHTQALLKVLQEQLKGTKELVETNGHSHEDSNEINVLTVALVNQDRENNIEKRSQ | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55068 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction