missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNOT8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA513451
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse based on sequence homology.
CNOT8 (CCR4-NOT transcription complex subunit 8), also known as CALIF or POP2, is a 292 amino acid protein that localizes to both the nucleus and the cytoplasm and functions as part of the CCR-NOT complex. Expressed ubiquitously, CNOT8 plays a role in transcriptional regulation for a diverse set of processes.
Specifications
| CNOT8 | |
| Polyclonal | |
| Unconjugated | |
| CNOT8 | |
| 1500015I04Rik; 1810022F04Rik; AA536816; AU015770; AU043059; CAF1; Caf1b; CAF1-like protein; CAF2; CALIF; CALIFp; CCR4-associated factor 8; CCR4-NOT transcription complex subunit 8; CCR4-NOT transcription complex, subunit 8; CNOT8; h CAF1-like protein; hCAF1; PGK promoter directed over production; POP2; wu:fe49a05; zgc:63844 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 9337 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9UFF9 | |
| CNOT8 | |
| KLH conjugated synthetic peptide between 227-255 amino acids from the C-terminal region of human CNOT8. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction