missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNOT8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568969
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
CNOT8 (CCR4-NOT transcription complex subunit 8), also known as CALIF or POP2, is a 292 amino acid protein that localizes to both the nucleus and the cytoplasm and functions as part of the CCR-NOT complex. Expressed ubiquitously, CNOT8 plays a role in transcriptional regulation for a diverse set of processes.
Specifications
| CNOT8 | |
| Polyclonal | |
| Unconjugated | |
| CNOT8 | |
| 1500015I04Rik; 1810022F04Rik; AA536816; AU015770; AU043059; CAF1; Caf1b; CAF1-like protein; CAF2; CALIF; CALIFp; CCR4-associated factor 8; CCR4-NOT transcription complex subunit 8; CCR4-NOT transcription complex, subunit 8; CNOT8; h CAF1-like protein; hCAF1; PGK promoter directed over production; POP2; wu:fe49a05; zgc:63844 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 69125 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9D8X5 | |
| CNOT8 | |
| synthetic peptide directed towards the middle region of human CNOT8 (aa 15-64). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction