missing translation for 'onlineSavingsMsg'
Learn More

CNOT7 Antibody (2F6), Novus Biologicals™

Product Code. 18367059 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367059 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367059 Supplier Novus Biologicals Supplier No. H00029883M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CNOT7 Monoclonal antibody specifically detects CNOT7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNOT7
Applications Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 2F6
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH60852
Gene Alias BTG1 binding factor 1, BTG1-binding factor 1, CAF-1, CAF1carbon catabolite repressor protein (CCR4)-associative factor 1, CCR4-associated factor 1, CCR4-NOT transcription complex subunit 7, CCR4-NOT transcription complex, subunit 7, hCAF-1
Host Species Mouse
Immunogen CNOT7 (AAH60852, 1 a.a. ~ 285 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPAATVDHSQRICEVWACNLDEEMKKIRQVIRKYNYVAMDTEFPGVVARPIGEFRSNADYQYQLLRCNVDLLKIIQLGLTFMNEQGEYPPGTSTWQFNFKFNLTEDMYAQDSIELLTTSGIQFKKHEEEGIETQYFAELLMTSGVVLCEGVKWLSFHSGYDFGYLIKILTNSNLPEEELDFFEILRLFFPVIYDVKYLMKSCKNLKGGLQEVAEQLELERIGPQHQAGSDSLLTGMAFFKMREMFFEDHIDDAKYCGHLYGLGSGSSYVQNGTGNAYEEEANKQS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29883
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.