Learn More
Invitrogen™ CNOT6L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114256
Description
Immunogen sequence: AKIMSEQERK HVDGCAIFFK TEKFTLVQKH TVEFNQVAMA NSDGSEAMLN For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
Has 3'-5' poly(A) exoribonuclease activity for synthetic poly(A) RNA substrate. Catalytic component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. May be involved in the deadenylation-dependent degradation of mRNAs through the 3'-UTR AU-rich element-mediated mechanism. Involved in deadenylation-dependent degradation of CDKN1B mRNA. Its mRNA deadenylase activity can be inhibited by TOB1. Mediates cell proliferation and cell survival and prevents cellular senescence. [UniProt].
Specifications
| CNOT6L | |
| Polyclonal | |
| Unconjugated | |
| CNOT6L | |
| Carbon catabolite repressor protein 4 homolog B; CCR4b; CCR4-NOT transcription complex subunit 6 like; CCR4-NOT transcription complex subunit 6-like; CCR4-NOT transcription complex, subunit 6-like; CNOT6L | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 246175 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96LI5 | |
| CNOT6L | |
| Synthetic peptide directed towards the middle region of Human CNOT6L. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.