Learn More
CNOT4 Mouse, anti-Human, CoraLite 594, Clone: 4H5H3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-67798-100UL
Description
CNOT4, also known as CCR4-NOT transcription complex subunit 4 , has E3 ubiquitin ligase activity, and promotes ubiquitination and degradation of target proteins. CNOT4 is involved in activation of the JAK/STAT pathway. CNOT4 exist many isoforms and the range of its molecular weight from 48 to 84 kDa.
Specifications
| CNOT4 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat |
| Immunofluorescence | |
| 4H5H3 | |
| CoraLite 594 | |
| 100 μL | |
| 4850 | |
| Liquid |