missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539800
This item is not returnable.
View return policy
Description
The CCR4-NOT complex functions as general transcription regulation complex. Subunit of the CCR4-NOT core complex that contains CHAF1A, CHAF1B, CNOT1, CNOT2, CNOT3, CNOT4, CNOT6 and CNOT8. 5 isoforms of the human protein are produced by alternative splicing.
Specifications
| CNOT2 | |
| Polyclonal | |
| Unconjugated | |
| CNOT2 | |
| NOT2H, CDC36, CNOT2, HSPC131 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 299805, 4848, 72068 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NZN8, Q8C5L3 | |
| CNOT2 | |
| A synthetic non-phosphopeptide derived from human CNOT2 around the phosphorylation site of Ser101 (S-L-SP-Q-G) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction