missing translation for 'onlineSavingsMsg'
Learn More

CNOT2 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18415934 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415934 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415934 Supplier Novus Biologicals Supplier No. NBP130454AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CNOT2 Polyclonal antibody specifically detects CNOT2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNOT2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CCR4-associated factor 2, CCR4-NOT transcription complex, subunit 2, CDC36FLJ26456, negative regulator of transcription 2, NOT2CCR4-NOT transcription complex subunit 2, NOT2H
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 490 and 540 of human CCR4-NOT transcription complex, subunit 2 using the numbering given in entry NP_055330.1 (GeneID 4848).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4848
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.