missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT10 Antibody (PCRP-CNOT10-1D5), Alexa Fluor™ 350, Novus Biologicals™
Shop All Bio Techne ProductsDescription
CNOT10 Monoclonal antibody specifically detects CNOT10 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | CNOT10 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | PCRP-CNOT10-1D5 |
| Conjugate | Alexa Fluor 350 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CCR4-NOT transcription complex subunit 10, CCR4-NOT transcription complex, subunit 10, DKFZp434K115, FLJ12890, FLJ13165 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human CNOT10 protein (Uniprot: Q9H9A5) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?