missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNOT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561253
This item is not returnable.
View return policy
Description
Immunogen sequence: TIETLMRINA HSRGNAPEGL PQLMEVVRSN YEAMIDRAHG GPNFMMHSGI SQASEYDDPP GLREKAEYLL REWVNLYHSA AAGRDSTKAF SAFVG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Scaffolding component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Its scaffolding function implies its interaction with the catalytic complex module and diverse RNA-binding proteins mediating the complex recruitment to selected mRNA 3'UTRs. Involved in degradation of AU-rich element (ARE)-containing mRNAs probably via association with ZFP36. Mediates the recruitment of the CCR4-NOT complex to miRNA targets and to the RISC complex via association with TNRC6A, TNRC6B or TNRC6C. Acts as a transcriptional repressor. Represses the ligand-dependent transcriptional activation by nuclear receptors. Involved in the maintenance of embryonic stem (ES) cell identity. [UniProt].
Specifications
| CNOT1 | |
| Polyclonal | |
| Unconjugated | |
| CNOT1 | |
| 6030411K04Rik; AA815922; AD-005; adrenal gland protein AD-005; CCR4-associated factor 1; CCR4-NOT transcription complex subunit 1; CCR4-NOT transcription complex, subunit 1; CDC39; Cnot1; D830048B13; Kiaa1007; Negative regulator of transcription subunit 1 homolog; NOT1; NOT1 (negative regulator of transcription 1, yeast) homolog; NOT1H; RGD1308009 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23019 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A5YKK6 | |
| CNOT1 | |
| Recombinant protein corresponding to Human CNOT1. Recombinant protein control fragment (Product #RP-95506). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction