missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92464-0.1ml
This item is not returnable.
View return policy
Description
CNOT1 Polyclonal antibody specifically detects CNOT1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| CNOT1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| AD-005, adrenal gland protein AD-005, CCR4-associated factor 1, CCR4-NOT transcription complex subunit 1, CCR4-NOT transcription complex, subunit 1, CDC39DKFZp686E0722, hNOT1, KIAA1007DKFZp686O168, Negative regulator of transcription subunit 1 homolog, NOT1 (negative regulator of transcription 1, yeast) homolog, NOT1FLJ90644, NOT1HFLJ36492 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 2137-2376 of human CNOT1 (NP_057368.3). PRNMRLPDPFTPNLKVDMLSEINIAPRILTNFTGVMPPQFKKDLDSYLKTRSPVTFLSDLRSNLQVSNEPGNRYNLQLINALVLYVGTQAIAHIHNKGSTPSMSTITHSAHMDIFQNLAVDLDTEGRYLFLNAIANQLRYPNSHTHYFSCTMLYLFAEANTEAIQEQITRVLLERLIVNRPHPWGLLITFIELIKNPAFKFWNHEFVHCAPEIEKLFQSVAQCCMGQKQAQQVMEGTGAS | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23019 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction