missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNNM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595885
This item is not returnable.
View return policy
Description
Immunogen sequence: VRALVDLQYI KITRQQYQNG LLASRMENSP QFPIDGCTTH MENLAEKSEL PVVDETTTLL NERNSLLHKA SHENAI.
This gene encodes a member of the ancient conserved domain containing protein family. Members of this protein family contain a cyclin box motif and have structural similarity to the cyclins. The encoded protein may play a role in metal ion transport. Mutations in this gene are associated with Jalili syndrome which consists of cone-rod dystrophy and amelogenesis imperfecta.
Specifications
| CNNM4 | |
| Polyclonal | |
| Unconjugated | |
| CNNM4 | |
| 5430430O18Rik; Acdp4; ancient conserved domain protein 4; ancient conserved domain-containing protein 4; Cnnm4; cyclin and CBS domain divalent metal cation transport mediator 4; cyclin M4; cyclin-M4; Kiaa1592; mACDP4; metal transporter CNNM4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26504, 363216, 94220 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.55 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P0C588, Q69ZF7, Q6P4Q7 | |
| CNNM4 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction