missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNNM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550216
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total Cyclin M3 protein.
CNNM3, also known as cyclin M3, is part of the CNNM family of proteins that play a critical role in magnesium ion transport across cellular membranes. This gene is notably expressed in the proximal tubule of the kidney, and its protein product localizes to the lateral membrane, suggesting its involvement in magnesium homeostasis. The structural analysis of the CNNM3 protein reveals it contains a cyclic nucleotide-binding homology domain with a characteristic beta-roll structure capped by an α-helical bundle, which is typical of cyclic nucleotide-binding domains. CNNM proteins were originally discovered in bacteria and are conserved across various organisms, indicating their fundamental biological importance. CNNM3 has been studied for its ability to decrease whole cellular magnesium content and free intracellular magnesium activity when expressed in model systems such as Xenopus laevis oocytes.
Specifications
| CNNM3 | |
| Polyclonal | |
| Unconjugated | |
| CNNM3 | |
| Acdp3; ancient conserved domain protein 3; ancient conserved domain-containing protein 3; Clp3; Cnnm3; cyclin and CBS domain divalent metal cation transport mediator 3; cyclin M3; cyclin-M3; mACDP3; metal transporter CNNM3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26505, 94218 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q32NY4, Q8NE01 | |
| CNNM3 | |
| Fusion protein corresponding to residues near the C terminal of human Cyclin M3. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction