Learn More
Invitrogen™ CNNM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120530
Description
Positive test controls include: Mouse brain, Rat brain. Immunogen sequence: NGAFTYYGVP AIMTTACSDN DVRKVGSLAG SSVFLNRSPS RCSGLNRSES PNRERSDFGG SNTQLYSSSN NLYMPDYSVH ILSDVQFVKI TRQQYQNALT ACHMDSSPQS PDMEAFTDGD STKAPTTRGT PQTPKDDPAI TLLNNRNSLP CSRSDGLRSP SEVVYLRMEE LAFTQEEMTD FEEHSTQQLT LSPAAVPTRA ASDSECCNIN LDTETSPCSS DFEENVGKKL LRTLSGQKRK RSPEGERTSE DNSNLTPLIT.
Probable metal transporter.
Specifications
| CNNM1 | |
| Polyclonal | |
| Unconjugated | |
| CNNM1 | |
| ACDP1; AI846314; ancient conserved domain protein 1; ancient conserved domain-containing protein 1; CLP-1; Cnnm1; cyclin and CBS domain divalent metal cation transport mediator 1; cyclin M1; cyclin-like protein 1; Cyclin-M1; mACDP1; Metal transporter CNNM1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26507, 309387, 83674 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.48 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q0GA42, Q9NRU3 | |
| CNNM1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 692-951 of human CNNM1 (NP_065081.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.