missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNKSR1 Mouse anti-Human, Clone: 1E11H8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CNKSR1 |
|---|---|
| Clone | 1E11H8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16809005
|
Proteintech
66796-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898995
|
Proteintech
66796-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a protein containing several motifs involved in protein-protein interaction, including PDZ, PH (Pleckstrin homology), and SAM (sterile alpha motif) domains. The encoded protein acts as a scaffold component for receptor tyrosine kinase signaling and may mediate crosstalk between different signaling pathways. Alternative splicing results in multiple transcript variants.Specifications
| CNKSR1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CNK, CNK homolog protein 1, CNK1, CNKSR1, Connector enhancer of KSR 1, Connector enhancer of KSR like, hCNK1, KSR | |
| CNKSR1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1E11H8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q969H4 | |
| 10256 | |
| CNKSR1 Fusion Protein Ag27855 | |
| Primary | |
| -20°C | |
| CNKSR1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title