missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNKSR1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97250AF647
This item is not returnable.
View return policy
Description
CNKSR1 Polyclonal antibody specifically detects CNKSR1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CNKSR1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 10256 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 647 | |
| CNK, CNK1, CNK1CNK homolog protein 1, connector enhancer of kinase suppressor of Ras 1, Connector enhancer of KSR 1, Connector enhancer of KSR-like, hCNK1, KSR | |
| Produced in rabbits immunized with E. coli-derived Human CNKSR1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction