missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNIH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551718
This item is not returnable.
View return policy
Description
Immunogen sequence: LLAYHIWRYM SRPVMSGPGL YDPTTIMNAD ILAYCQKEGW CKL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
CNIH is involved in the selective transport and maturation of TGF-alpha family proteins.
Specifications
| CNIH1 | |
| Polyclonal | |
| Unconjugated | |
| CNIH1 | |
| 0610007J15; Cnih; CNIH1; CNIH-1; CNIL; Cornichon family AMPA receptor auxiliary protein 1; cornichon homolog; cornichon homolog 1 (Drosophila); cornichon-like isoform 1; hypothetical protein LOC613143; protein cornichon homolog; Protein cornichon homolog 1; T-cell growth-associated molecule 77; TGAM77; UNQ155/PRO181; wu:fc21b06; wu:fi12c05; zgc:112228; zgc:56103 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10175 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95406 | |
| CNIH1 | |
| Recombinant protein corresponding to Human CNIH1. Recombinant protein control fragment (Product #RP-89079). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction