missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNIH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98134-50ul
This item is not returnable.
View return policy
Description
CNIH Polyclonal antibody specifically detects CNIH in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CNIH | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| CNIH1, CNILMGC117156, cornichon homolog (Drosophila), T-cell growth-associated molecule 77, TGAM77protein cornichon homolog | |
| Produced in rabbits immunized with E. coli-derived Human CNIH fragment. | |
| 50 μg | |
| Signal Transduction | |
| 10175 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction