missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNGB3 Polyclonal Antibody

Brand: Thermo Scientific PA5114294
This item is not returnable.
View return policy
Description
Specifications
| CNGB3 | |
| Polyclonal | |
| Unconjugated | |
| CNGB3 | |
| ACHM1; cng channel; CNG channel beta-3; CNGB3; cone photoreceptor cGMP-gated cation channel beta-subunit; Cone photoreceptor cGMP-gated channel subunit beta; cyclic nucleotide gated channel beta 3; cyclic nucleotide-gated cation channel beta-3; Cyclic nucleotide-gated cation channel modulatory subunit; Cyclic nucleotide-gated channel beta-3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 54714 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NQW8 | |
| CNGB3 | |
| Synthetic peptide directed towards the N-terminal region of Human CNGB3 (aa 84-133). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction