missing translation for 'onlineSavingsMsg'
Learn More

CNGB3 Antibody, Novus Biologicals™

Product Code. 18396081 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396081 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396081 Supplier Novus Biologicals Supplier No. NBP275087

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

CNGB3 Polyclonal antibody specifically detects CNGB3 in Human, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNGB3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ELISA
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry 2-4 μg/mL, Immunohistochemistry-Paraffin 2-4 μg/mL, Immunohistochemistry-Frozen, Peptide ELISA 1:8000
Formulation TBS (pH 7.3), 0.5% BSA with 0.02% Sodium Azide
Gene Alias cyclic nucleotide gated channel beta 3
Gene Symbols CNGB3
Host Species Goat
Immunogen Peptide with sequence C-ESDDKPTEHYYR, from the internal region of the protein sequence according to NP_061971.3.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54714.0
Target Species Human, Monkey
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.